2,4-Dimethoxy-beta-methylcinnamic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

D5047-1G D5047-5G
76219-806EA 102.7 CAD
76219-806 76219-808
2,4-Dimethoxy-beta-methylcinnamic acid ≥98.0% (by GC, titration analysis)
2,4-Dimethoxy-beta-methylcinnamic acid
Formula: C₁₂H₁₄O₄
MDL Number: MFCD00512029
CAS Number: 7706-67-4
Merck Index: 14,03203

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 156.0 to 160.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR