2-Benzoylpyrrole ≥98.0% (by GC)

Supplier: TCI America
Warning

B4934-25G B4934-5G
TCB4934-25GEA 859.52 CAD
TCB4934-25G TCB4934-5G
2-Benzoylpyrrole ≥98.0% (by GC)
2-Benzoylpyrrole
Formula: C₁₁H₉NO
Boiling Pt: 305°C
MDL Number: MFCD00094048
CAS Number: 7697-46-3

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 75.0 to 79.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR