2,5-Dichloro-N-[2-(isopropylsulfonyl)phenyl]pyrimidin-4-amine ≥97.0% (by HPLC, total nitrogen)

Supplier: TCI America

D5068-1G D5068-200MG
76219-824EA 342.86 CAD
76219-824 76219-826
2,5-Dichloro-N-[2-(isopropylsulfonyl)phenyl]pyrimidin-4-amine ≥97.0% (by HPLC, total nitrogen)
2,5-Dichloro-N-[2-(isopropylsulfonyl)phenyl]pyrimidin-4-amine
Formula: C₁₃H₁₃Cl₂N₃O₂S
MDL Number: MFCD12827956
CAS Number: 761440-16-8

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 97.0 %
Melting point 149.0 to 153.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR