2-(4-Iodophenyl)-1-phenylbenzimidazole ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

I0983-1G I0983-200MG
76219-988EA 391.84 CAD
76219-988 76219-990
2-(4-Iodophenyl)-1-phenylbenzimidazole ≥98.0% (by HPLC, titration analysis)
2-(4-Iodophenyl)-1-phenylbenzimidazole
Formula: C₁₉H₁₃IN₂
MDL Number: MFCD29089366
CAS Number: 760212-42-8

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 178.0 to 182.0 deg-C

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR