1,3-Dibromo-5-hexylbenzene ≥95.0% (by GC)
Supplier: TCI America
|
Warning
|
D4789-1G
D4789-5G
TCD4789-1GEA
96.38
CAD
TCD4789-1G
TCD4789-5G
1,3-Dibromo-5-hexylbenzene ≥95.0% (by GC)
1,3-Dibromo-5-hexylbenzene
Formula:
C₁₂H₁₆Br₂ |
CAS Number:
75894-97-2 |
Specification Test Results
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...