2,6-Diethynylpyridine ≥98.0% (by GC)

Supplier: TCI America
Warning

D5097-1G D5097-200MG
76219-150EA 461.36 CAD
76219-150 76219-152
2,6-Diethynylpyridine ≥98.0% (by GC)
2,6-Diethynylpyridine
Formula: C₉H₅N
Storage Temperature: Refrigerator
MDL Number: MFCD09039127
CAS Number: 75867-46-8

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystaline
Purity (GC) min. 98.0 %
Melting point 69.0 to 73.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR