4-(4-Amino-3-fluorophenoxy)-N-methyl-2-pyridinecarboxamide ≥98.0% (by HPLC)

Supplier: TCI America
Warning

A2979-1G A2979-200MG
TCA2979-1GEA 158 CAD
TCA2979-1G TCA2979-200MG
4-(4-Amino-3-fluorophenoxy)-N-methyl-2-pyridinecarboxamide ≥98.0% (by HPLC)
4-(4-Amino-3-fluorophenoxy)-N-methyl-2-pyridinecarboxamide
Formula: C₁₃H₁₂FN₃O₂
MDL Number: MFCD09033852
CAS Number: 757251-39-1

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 95.0 %
Melting point 141.0 to 145.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR