Moxonidine ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Danger

M2660-1G M2660-200MG
TCM2660-1GEA 643.06 CAD
TCM2660-1G TCM2660-200MG
Moxonidine ≥98.0% (by titrimetric analysis)
Moxonidine
Formula: C₉H₁₂ClN₅O
MDL Number: MFCD00865794
CAS Number: 75438-57-2
UN: 2811
ADR: 6.1,III
Merck Index: 14,06293

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR