Monomethyl 1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
M2493-25G
M2493-5G
TCM2493-25GEA
173.8
CAD
TCM2493-25G
TCM2493-5G
Monomethyl 1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylate ≥98.0% (by HPLC, titration analysis)
Monomethyl 1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylate
Formula:
C₁₆H₁₆N₂O₆ |
MDL Number:
MFCD00475478 CAS Number: 74936-72-4 EINECS: 278-034-2 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 201.0 to 205.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...