Ethyl 2-Iodo-2-methylpropionate ≥94.0% (by GC)

Supplier: TCI America
Warning

E1199-25G E1199-5G
TCE1199-25GEA 417.12 CAD
TCE1199-25G TCE1199-5G
Ethyl 2-Iodo-2-methylpropionate ≥94.0% (by GC)
Ethyl 2-Iodo-2-methylpropionate
Formula: C₆H₁₁IO₂
Storage Temperature: Freezer
MDL Number: MFCD29089363
CAS Number: 7425-55-0

Order Now

Specification Test Results

Appearance Light yellow to Brown clear liquid
Purity (GC) min. 94.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR