9-(2-Naphthyl)anthracene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 9-(Naphthalen-2-yl)anthracene

N1130-1G N1130-5G
TCN1130-1GEA 107.44 CAD
TCN1130-1G TCN1130-5G
9-(2-Naphthyl)anthracene ≥98.0% (by GC)
9-(2-Naphthyl)anthracene
Formula: C₂₄H₁₆
MW: 304.39 g/mol
MDL Number: MFCD18205835
CAS Number: 7424-72-8

Order Now

Specification Test Results

Appearance White to Light yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 203.0 to 207.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR