9-(1-Naphthyl)anthracene ≥98.0% (by GC)

Supplier: TCI America

N0994-1G N0994-5G
TCN0994-1GEA 231.47 CAD
TCN0994-1G TCN0994-5G
9-(1-Naphthyl)anthracene ≥98.0% (by GC)
9-(1-Naphthyl)anthracene
Formula: C₂₄H₁₆
MDL Number: MFCD19689379
CAS Number: 7424-70-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 163.0 to 166.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR