3,6-Dibromo-9-(4-tert-butylphenyl)-9H-carbazole ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
|
Warning
|
D5000-1G
D5000-200MG
TCD5000-1GEA
240.16
CAD
TCD5000-1G
TCD5000-200MG
3,6-Dibromo-9-(4-tert-butylphenyl)-9H-carbazole ≥98.0% (by HPLC, total nitrogen)
3,6-Dibromo-9-(4-tert-butylphenyl)-9H-carbazole
Formula:
C₂₂H₁₉Br₂N |
MDL Number:
MFCD12024283 CAS Number: 741293-42-5 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 216.0 to 220.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...