Isobutylamine hydrobromide ≥98.0% (by total nitrogen and titration analysis)

Supplier: TCI America
Warning

I1007-1G I1007-5G
TCI1007-1GEA 66.36 CAD
TCI1007-1G TCI1007-5G
Isobutylamine hydrobromide ≥98.0% (by total nitrogen and titration analysis)
Isobutylamine hydrobromide
Formula: C₄H₁₁N·HBr
MDL Number: MFCD29089367
CAS Number: 74098-36-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR