Ketanserin ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 3-[2-[4-(4-Fluorobenzoyl)-1-piperidinyl]ethyl]-2,4(1H,3H)-quinazolinedione, 3-(2-(4-(4-Fluorobenzoyl)piperidin-1-yl)ethyl)quinazoline-2,4(1H,3H)-dione

K0051-1G K0051-200MG
TCK0051-1GEA 295.46 CAD
TCK0051-1G TCK0051-200MG
Ketanserin ≥98.0% (by HPLC, titration analysis)
Ketanserin
Formula: C₂₂H₂₂FN₃O₃
MW: 395.43 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00083392
CAS Number: 74050-98-9
EINECS: 277-680-2
Merck Index: 14,05296

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 231.0 to 235.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR