N-Benzyl-2,2,2-trifluoroacetamide ≥95.0% (by GC)

Supplier: TCI America

B5705-1G B5705-5G
TCB5705-1GEA 69.52 CAD
TCB5705-1G TCB5705-5G
N-Benzyl-2,2,2-trifluoroacetamide ≥95.0% (by GC)
N-Benzyl-2,2,2-trifluoroacetamide
Formula: C₉H₈F₃NO
MDL Number: MFCD00188542
CAS Number: 7387-69-1

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 95.0 %
Melting point 70.0 to 74.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR