10-Hexylphenothiazine ≥98.0% (by GC)

Supplier: TCI America
Warning

H1608-1G H1608-5G
76220-144EA 241.74 CAD
76220-144 76220-146
10-Hexylphenothiazine ≥98.0% (by GC)
10-Hexylphenothiazine
Formula: C₁₈H₂₁NS
CAS Number: 73025-93-1

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR