2-Dodecylhexadecan-1-ol ≥93.0% (by GC)

Supplier: TCI America

D5322-250G D5322-25G
TCD5322-250GEA 325.48 CAD
TCD5322-250G TCD5322-25G
2-Dodecylhexadecan-1-ol ≥93.0% (by GC)
2-Dodecylhexadecan-1-ol
Formula: C₂₈H₅₈O
Storage Temperature: Refrigerator
MDL Number: MFCD27665210
CAS Number: 72388-18-2
EINECS: 276-627-0

Order Now

Specification Test Results

Appearance White to Almost white powder to lump
Purity (GC) min. 93.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR