5-Methyl-4,5,6,7-tetrahydrothiazolo[5,4-c]pyridine-2-carboxylic acid hydrochloride ≥97.0% (by HPLC)

Supplier: TCI America

M2819-1G M2819-200MG
TCM2819-1GEA 112.18 CAD
TCM2819-1G TCM2819-200MG
5-Methyl-4,5,6,7-tetrahydrothiazolo[5,4-c]pyridine-2-carboxylic acid hydrochloride ≥97.0% (by HPLC)
5-Methyl-4,5,6,7-tetrahydrothiazolo[5,4-c]pyridine-2-carboxylic acid hydrochloride
Formula: C₈H₁₀N₂O₂S·HCl
MDL Number: MFCD25542379
CAS Number: 720720-96-7

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 94.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR