Isopropamide iodide ≥95.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

I1048-1G I1048-5G
TCI1048-1GEA 53.72 CAD
TCI1048-1G TCI1048-5G
Isopropamide iodide ≥95.0% (by HPLC, titration analysis)
Isopropamide iodide
Formula: C₂₃H₃₃IN₂O
Melting Pt: 201°C
MDL Number: MFCD00063513
CAS Number: 71-81-8
EINECS: 200-766-8
Merck Index: 14,05202

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Nonaqueous Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR