4-Bromo-9,9-diphenylfluorene ≥98.0% (by GC)

Supplier: TCI America

B4776-1G B4776-5G
TCB4776-1GEA 95.91 CAD
TCB4776-1G TCB4776-5G
4-Bromo-9,9-diphenylfluorene ≥98.0% (by GC)
4-Bromo-9,9-diphenylfluorene
Formula: C₂₅H₁₇Br
MDL Number: MFCD27923067
CAS Number: 713125-22-5

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 213.0 to 217.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR