2-(9-Anthryl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥98.0% (by GC)

Supplier: TCI America

A3184-1G A3184-5G
76219-320EA 104.28 CAD
76219-320 76219-322
2-(9-Anthryl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥98.0% (by GC)
2-(9-Anthryl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formula: C₂₀H₂₁BO₂
MDL Number: MFCD24039741
CAS Number: 709022-63-9

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 139.0 to 143.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR