2-[(5'-Bromo-[2,2'-bithiophen]-5-yl)methylene]malononitrile ≥95.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 2-[(5'-Bromo-[2,2'-bithiophen]-5-yl)methylene]propanedinitrile

B5458-1G B5458-200MG
76219-618EA 485.06 CAD
76219-618 76219-620
2-[(5'-Bromo-[2,2'-bithiophen]-5-yl)methylene]malononitrile ≥95.0% (by GC)
2-[(5'-Bromo-[2,2'-bithiophen]-5-yl)methylene]malononitrile
Formula: C₁₂H₅BrN₂S₂
MDL Number: MFCD06214120
CAS Number: 704890-84-6

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 95.0 %
Melting point 200.0 to 204.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR