Phenyl trifluoromethyl sulfoxide ≥98.0% (by GC)

Supplier: TCI America
Warning

P2460-1G P2460-5G
TCP2460-1GEA 230.68 CAD
TCP2460-1G TCP2460-5G
Phenyl trifluoromethyl sulfoxide ≥98.0% (by GC)
Phenyl trifluoromethyl sulfoxide
Formula: C₇H₅F₃OS
Boiling Pt: 82°C
MDL Number: MFCD30475661
CAS Number: 703-18-4

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR