4'-Cyano-4-decyloxybiphenyl ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 10OCB, 4'-Decyloxy-4-biphenylcarbonitrile, 4'-(Decyloxy)-[1,1'-biphenyl]-4-carbonitrile

C3269-1G C3269-5G
TCC3269-1GEA 285.98 CAD
TCC3269-1G TCC3269-5G
4'-Cyano-4-decyloxybiphenyl ≥98.0% (by GC)
4'-Cyano-4-decyloxybiphenyl
Formula: C₂₃H₂₉NO
MDL Number: MFCD01218034
CAS Number: 70247-25-5
EINECS: 274-495-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Mesomorphic range 79.0 to 87.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR