N-Succinimidyl tetradecanoate ≥95.0% (by HPLC, total nitrogen)
Supplier: TCI America
|
Warning
|
S0997-1G
S0997-5G
TCS0997-1GEA
271.76
CAD
TCS0997-1G
TCS0997-5G
N-Succinimidyl tetradecanoate ≥95.0% (by HPLC, total nitrogen)
N-Succinimidyl tetradecanoate
Formula:
C₁₈H₃₁NO₄ Storage Temperature: Freezer |
MDL Number:
MFCD00522592 CAS Number: 69888-86-4 |
Specification Test Results
Purity (HPLC) | min. 95.0 area% |
Purity (with Total Nitrogen) | min. 95.0 % |
Melting point | 82.0 to 86.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...