N,N-Diphenyl-2-naphthylamine ≥98.0% (by GC)

Supplier: TCI America
Warning

D4843-1G D4843-5G
TCD4843-1GEA 224.36 CAD
TCD4843-1G TCD4843-5G
N,N-Diphenyl-2-naphthylamine ≥98.0% (by GC)
N,N-Diphenyl-2-naphthylamine
Formula: C₂₂H₁₇N
MDL Number: MFCD28384144
CAS Number: 6940-30-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 123.0 to 127.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR