2-Methoxycarbazole ≥98.0% (by GC)

Supplier: TCI America
Warning

M2817-1G M2817-5G
TCM2817-1GEA 300.2 CAD
TCM2817-1G TCM2817-5G
2-Methoxycarbazole ≥98.0% (by GC)
2-Methoxycarbazole
Formula: C₁₃H₁₁NO
MDL Number: MFCD09033506
CAS Number: 6933-49-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 237.0 to 241.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR