Germacrone ≥98.0% (by GC)

Supplier: TCI America
Warning

G0519-100MG G0519-20MG
TCG0519-100MGEA 799.48 CAD
TCG0519-100MG TCG0519-20MG
Germacrone ≥98.0% (by GC)
Germacrone
Formula: C₁₅H₂₂O
Storage Temperature: Refrigerator
MDL Number: MFCD00210050
CAS Number: 6902-91-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 54.0 to 58.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR