Calcium(II) bis(nonafluorobutanesulfonyl)imide ≥95.0% (by titrimetric analysis)

Supplier: TCI America
Danger

C3447-1G C3447-5G
TCC3447-1GEA 145.36 CAD
TCC3447-1G TCC3447-5G
Calcium(II) bis(nonafluorobutanesulfonyl)imide ≥95.0% (by titrimetric analysis)
Calcium(II) bis(nonafluorobutanesulfonyl)imide
Formula: C₁₆CaF₃₆N₂O₈S₄
CAS Number: 689282-13-1
UN: 1759
ADR: 8,II

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Ion exchange titration) min. 95.0 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR