4,4'-Dibromo-1,1'-spirobiindane-7,7'-diol ≥98.0% (by GC)
Supplier: TCI America
D4332-1G
D4332-5G
TCD4332-1GEA
323.9
CAD
TCD4332-1G
TCD4332-5G
4,4'-Dibromo-1,1'-spirobiindane-7,7'-diol ≥98.0% (by GC)
4,4'-Dibromo-1,1'-spirobiindane-7,7'-diol
Formula:
C₁₇H₁₄Br₂O₂ |
MDL Number:
MFCD16660947 CAS Number: 681481-91-4 |
Specification Test Results
Purity (GC) | min. 98.0 % |
Melting point | 146.0 to 150.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...