Imidazole Hydroiodide, low water content ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: Imidazolium iodide

I0970-1G I0970-5G
TCI0970-1GEA 124.03 CAD
TCI0970-1G TCI0970-5G
Imidazole Hydroiodide, low water content ≥98.0% (by titrimetric analysis)
Imidazole Hydroiodide
Formula: C₃H₄N₂·HI
MW: 195.99 g/mol
CAS Number: 68007-08-9

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Water max. 100 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR