cis-4-Nitrostilbene ≥97.0% (by GC)

Supplier: TCI America

N1085-1G N1085-200MG
TCN1085-1GEA 709.42 CAD
TCN1085-1G TCN1085-200MG
cis-4-Nitrostilbene ≥97.0% (by GC)
cis-4-Nitrostilbene
Formula: C₁₄H₁₁NO₂
CAS Number: 6624-53-9

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Green powder to crystal
Purity (GC) min. 97.0 %
Melting point 61.0 to 65.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR