Dienogest ≥98.0% (by HPLC)

Supplier: TCI America
Warning

D5230-250MG D5230-50MG
TCD5230-250MGEA 944.84 CAD
TCD5230-250MG TCD5230-50MG
Dienogest ≥98.0% (by HPLC)
Dienogest
Formula: C₂₀H₂₅NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD00868356
CAS Number: 65928-58-7
Merck Index: 14,03106

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 4.20 to 4.80 %
Melting point 213.0 to 217.0 deg-C
Specific rotation [a]20/D -302 to -312 deg (C=1, CH2Cl2)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR