4-Ethyl-4-methylmorpholinium bromide ≥97.0% (by titrimetric analysis)

Supplier: TCI America
Warning

E1166-25G E1166-5G
TCE1166-25GEA 167.48 CAD
TCE1166-25G TCE1166-5G
4-Ethyl-4-methylmorpholinium bromide ≥97.0% (by titrimetric analysis)
4-Ethyl-4-methylmorpholinium bromide
Formula: C₇H₁₆BrNO
MDL Number: MFCD03095385
CAS Number: 65756-41-4
EINECS: 418-250-8

Order Now

Specification Test Results

Appearance White to Light yellow to Dark green powder to crystal
Purity (Nonaqueous Titration) min. 97.0 %
Purity (Argentmetric Titration) min. 97.0 %
Melting point 177.0 to 187.0 deg-C

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR