2,2,2-Trifluoro-3'-nitroacetophenone ≥98.0% (by GC)

Supplier: TCI America
Warning

T3426-1G T3426-5G
76220-442EA 150.1 CAD
76220-442 76220-624
2,2,2-Trifluoro-3'-nitroacetophenone ≥98.0% (by GC)
2,2,2-Trifluoro-3'-nitroacetophenone
Formula: C₈H₄F₃NO₃
Boiling Pt: 113°C
MDL Number: MFCD00034541
CAS Number: 657-15-8
EINECS: 211-516-2

Order Now

Specification Test Results

Appearance White to Light yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 54.0 to 58.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR