(1R,2R)-1,2-Cyclohexanedimethanol ≥98.0% (by GC)

Supplier: TCI America

C2978-1G C2978-5G
TCC2978-1GEA 202.24 CAD
TCC2978-1G TCC2978-5G
(1R,2R)-1,2-Cyclohexanedimethanol ≥98.0% (by GC)
(1R,2R)-1,2-Cyclohexanedimethanol
Formula: C₈H₁₆O₂
Boiling Pt: 113°C
MDL Number: MFCD16660860
CAS Number: 65376-05-8

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 63.0 to 67.0 deg-C
Specific rotation [a]20/D +16.0 to +20.0 deg (C=1, toluene)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR