4-Iododibenzofuran ≥98.0% (by GC)
Supplier: TCI America
I0972-1G
I0972-200MG
TCI0972-1GEA
818.12
CAD
TCI0972-1G
TCI0972-200MG
4-Iododibenzofuran ≥98.0% (by GC)
4-Iododibenzofuran
Formula:
C₁₂H₇IO Melting Pt: 73°C |
MDL Number:
MFCD00094364 CAS Number: 65344-26-5 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...