Methyl (3,5-Dimethoxyphenyl)acetate ≥98.0% (by GC)

Supplier: TCI America

M2933-1G M2933-5G
TCM2933-1GEA 132.72 CAD
TCM2933-1G TCM2933-5G
Methyl (3,5-Dimethoxyphenyl)acetate ≥98.0% (by GC)
Methyl (3,5-Dimethoxyphenyl)acetate
Formula: C₁₁H₁₄O₄
Boiling Pt: 94°C
MDL Number: MFCD00089971
CAS Number: 6512-32-9

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR