Ethyl (Z)-2-(2-Amino-4-thiazolyl)-2-(methoxyimino)acetate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
Ethyl (Z)-2-Amino-alpha-(methoxyimino)-4-thiazoleacetate, (Z)-2-Amino-alpha-(methoxyimino)-4-thiazoleacetic acid Ethyl ester, (Z)-2-(2-Amino-4-thiazolyl)-2-(methoxyimino)acetic acid Ethyl ester
E0990-25G
E0990-5G
TCE0990-25GEA
323.9
CAD
TCE0990-25G
TCE0990-5G
Ethyl (Z)-2-(2-Amino-4-thiazolyl)-2-(methoxyimino)acetate ≥98.0% (by HPLC, titration analysis)
Ethyl (Z)-2-(2-Amino-4-thiazolyl)-2-(methoxyimino)acetate
Formula:
C₈H₁₁N₃O₃S MW: 229.25 g/mol |
MDL Number:
MFCD00071532 CAS Number: 64485-88-7 EINECS: 264-912-2 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 162.0 to 166.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...