3,3,3',3'-Tetramethyl-1,1'-bis(4-sulfobutyl)benzoindodicarbocyanine sodium salt 98,0 HPLC, N

Supplier: TCI America
Warning

Synonyms: 2-[5-[1,1-Dimethyl-3-(4-sulfobutyl)-1,3-dihydrobenzo[e]indol-2-ylidene]-1,3-pentadienyl]-1,1-dimethyl-3-(4-sulfobutyl)-1H-benzo[e]indolium hydroxide Inner salt sodium salt, Sodium 4-[2-[5-[1,1-Dimethyl-3-(4-sulphonatobutyl)-1H-benz[e]indol-2(3H)-ylidene]-1,3-pentadienyl]-1,1-dimethyl-1H-benz[e]indol-3-ium-3-yl]butane-1-sulphonate, Sodium 4-[2-[5-[1,1-Dimethyl-3-(4-sulfonatobutyl)-1H-benz[e]indol-2(3H)-ylidene]-1,3-pentadienyl]-1,1-dimethyl-1H-benz[e]indol-3-ium-3-yl]butane-1-sulfonate, 5-[1,1-Dimethyl-3-(4-sulfonatobutyl)benz[e]indolin-2-ylidene]-1-[1,1-dimethyl-3-(4-sulfonatobutyl)benz[e]indolium-2-yl]-1,3-pentadiene sodium salt, NK 1841, 5-[1,1-Dimethyl-3-(4-sulphonatobutyl)benz[e]indolin-2-ylidene]-1-[1,1-dimethyl-3-(4-sulphonatobutyl)benz[e]indolium-2-yl]-1,3-pentadiene sodium salt, 2-[5-[1,1-Dimethyl-3-(4-sulphobutyl)-1,3-dihydrobenzo[e]indol-2-ylidene]-1,3-pentadienyl]-1,1-dimethyl-3-(4-sulphobutyl)-1H-benzo[e]indolium hydroxide Inner salt sodium salt

D4463-1G D4463-5G
TCD4463-1GEA 271.76 CAD
TCD4463-1G TCD4463-5G
3,3,3',3'-Tetramethyl-1,1'-bis(4-sulfobutyl)benzoindodicarbocyanine sodium salt 98,0 HPLC, N
3,3,3',3'-Tetramethyl-1,1'-bis(4-sulfobutyl)benzoindodicarbocyanine sodium salt
Formula: C₄₁H₄₅N₂NaO₆S₂
MW: 748.93 g/mol
MDL Number: MFCD16621036
CAS Number: 64285-36-5

Order Now

Specification Test Results

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Absorbance (E1%1cm) min. 2800 (MeOH, 675 to 685nm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR