2-(4-Bromophenyl)pyridine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
B4500-1G
B4500-5G
TCB4500-1GEA
99.54
CAD
TCB4500-1G
TCB4500-5G
2-(4-Bromophenyl)pyridine ≥98.0% (by GC, titration analysis)
2-(4-Bromophenyl)pyridine
Formula:
C₁₁H₈BrN |
MDL Number:
MFCD04116231 CAS Number: 63996-36-1 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 61.0 to 65.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...