5-Chloro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene ≥97.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 5-Chloro-2-thiopheneboronic acid Pinacol ester, 2-(5-Chloro-2-thienyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

C3301-1G C3301-5G
TCC3301-1GEA 139.04 CAD
TCC3301-1G TCC3301-5G
5-Chloro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene ≥97.0% (by GC, titration analysis)
5-Chloro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene
Formula: C₁₀H₁₄BClO₂S
MW: 244.54 g/mol
MDL Number: MFCD08063140
CAS Number: 635305-24-7

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR