(R)-(-)-alpha-[[(4-Ethyl-2,3-dioxo-1-piperazinyl)carbonyl]amino]benzeneacetic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

E0993-1G E0993-5G
TCE0993-1GEA 45.82 CAD
TCE0993-1G TCE0993-5G
(R)-(-)-alpha-[[(4-Ethyl-2,3-dioxo-1-piperazinyl)carbonyl]amino]benzeneacetic acid ≥98.0% (by HPLC, titration analysis)
(R)-(-)-alpha-[[(4-Ethyl-2,3-dioxo-1-piperazinyl)carbonyl]amino]benzeneacetic acid
Formula: C₁₅H₁₇N₃O₅
MDL Number: MFCD00799536
CAS Number: 63422-71-9
EINECS: 264-133-8

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D -64.0 to -68.0 deg (C=1,MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR