1-Chloro-2-naphthol ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

C3175-1G C3175-5G
76218-914EA 115.34 CAD
76218-914 76218-916
1-Chloro-2-naphthol ≥98.0% (by GC, titration analysis)
1-Chloro-2-naphthol
Formula: C₁₀H₇ClO
Boiling Pt: 157°C
MDL Number: MFCD00997084
CAS Number: 633-99-8
EINECS: 211-200-4

Order Now

Specification Test Results

Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 66.0 to 70.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR