(4-Allyl-2-methoxyphenoxy)acetic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

A3037-1G A3037-5G
76218-782EA 48.98 CAD
76218-782 76218-784
(4-Allyl-2-methoxyphenoxy)acetic acid ≥98.0% (by GC, titration analysis)
(4-Allyl-2-methoxyphenoxy)acetic acid
Formula: C₁₂H₁₄O₄
MDL Number: MFCD00017594
CAS Number: 6331-61-9

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 99.0 to 103.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR