N-(1-Naphthyl)formamide ≥98.0% (by GC)
Supplier: TCI America
N1001-25G
N1001-5G
TCN1001-25GEA
529.3
CAD
TCN1001-25G
TCN1001-5G
N-(1-Naphthyl)formamide ≥98.0% (by GC)
N-(1-Naphthyl)formamide
Formula:
C₁₁H₉NO MW: 171.20 g/mol |
MDL Number:
MFCD00086455 CAS Number: 6330-51-4 |
Specification Test Results
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 138.0 to 142.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...