3-Acetamidophthalic anhydride ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 3-Acetamidophthalic Anhydride, N-(1,3-Dioxo-1,3-dihydroisobenzofuran-4-yl)acetamide

A2804-1G A2804-5G
TCA2804-1GEA 60.04 CAD
TCA2804-1G TCA2804-5G
3-Acetamidophthalic anhydride ≥98.0% (by GC, titration analysis)
3-Acetamidophthalic anhydride
Formula: C₁₀H₇NO₄
MW: 205.17 g/mol
MDL Number: MFCD00453138
CAS Number: 6296-53-3

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 99.0 %
Purity (Methanolysis Method) min. 98.0 %
Melting point 184.0 to 188.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR