(R)-(-)-alpha-[[(4-Ethyl-2,3-dioxo-1-piperazinyl)carbonyl]amino]-4-hydroxybenzeneacetic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

E0994-1G E0994-5G
TCE0994-1GEA 37.92 CAD
TCE0994-1G TCE0994-5G
(R)-(-)-alpha-[[(4-Ethyl-2,3-dioxo-1-piperazinyl)carbonyl]amino]-4-hydroxybenzeneacetic acid ≥98.0% (by HPLC, titration analysis)
(R)-(-)-alpha-[[(4-Ethyl-2,3-dioxo-1-piperazinyl)carbonyl]amino]-4-hydroxybenzeneacetic acid
Formula: C₁₅H₁₇N₃O₆
MDL Number: MFCD00792472
CAS Number: 62893-24-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D -84.0 to -87.0 deg (C=1,MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR