9-Hexyl-3,6-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

H1649-1G H1649-5G
TCH1649-1GEA 211.72 CAD
TCH1649-1G TCH1649-5G
9-Hexyl-3,6-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole ≥98.0% (by HPLC, total nitrogen)
9-Hexyl-3,6-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole
Formula: C₃₀H₄₃B₂NO₄
MDL Number: MFCD20528097
CAS Number: 628336-95-8

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 155.0 to 159.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR